Learn More
MSC Kwazar Venus Pro+ 360 Handheld Sprayer, 1.5 L, High Density Polyethylene, Viton Seals
Catalog No. 502838401
Encompass
The Kwazar Venus Pro+ 360 Sprayer is a compression/pump-up sprayer designed for ease of use. Its 360-degree feature allows continuous spraying from various angles, including inverted positions, ensuring full product utilization. It features seals resistant to common chemicals, a pressure release safety valve, and a movable nozzle for varied spray patterns. An embossed level indicator shows measurements in milliliters and pints. Ideal for car care, detailing, and professional cleaning.
  • Easy-to-use compression/pump-up design.
  • 360-degree functionality for continuous spraying at all angles.
  • FPM seals resist common chemicals and detergents.
  • Includes a pressure release safety valve.
  • Movable nozzle for varied spray patterns.
  • Embossed level indicator in milliliters and pints.
  • Features an in-tank filter.
  • Adjustable nozzle included.
Catalog No. 50-283-8401 Supplier MSC Supplier No. 74255993
The Kwazar Venus Pro+ 360 Sprayer is a compression/pump-up sprayer designed for ease of use. Its 360-degree feature allows continuous spraying from various angles, including inverted positions, ensuring full product utilization. It features seals resistant to common chemicals, a pressure release safety valve, and a movable nozzle for varied spray patterns. An embossed level indicator shows measurements in milliliters and pints. Ideal for car care, detailing, and professional cleaning.
  • Easy-to-use compression/pump-up design.
  • 360-degree functionality for continuous spraying at all angles.
  • FPM seals resist common chemicals and detergents.
  • Includes a pressure release safety valve.
  • Movable nozzle for varied spray patterns.
  • Embossed level indicator in milliliters and pints.
  • Features an in-tank filter.
  • Adjustable nozzle included.