Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Sorbent, 10g/Vial

Catalog No. 502393297 Shop All Phenomenex Products
Encompass

Venusil 10m PrepG C18 120 VX900010-A

Catalog No. 50-239-3297 Supplier Phenomenex Inc Supplier No. VX900010A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 10m PrepG C18 120 VX900010-A

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.