Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 150 x 21.2mm

Catalog No. 502395117 Shop All Phenomenex Products
Encompass

Venusil 10m PrepG C18 120 VX901520-A

Catalog No. 50-239-5117 Supplier Phenomenex Inc Supplier No. VX901520A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 10m PrepG C18 120 VX901520-A

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.