Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 100 x 4mm

Catalog No. 502394942 Shop All Phenomenex Products
Encompass

Venusil 3m XBP C18 100 VX931004-0

Catalog No. 50-239-4942 Supplier Phenomenex Inc Supplier No. VX9310040
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 3m XBP C18 100 VX931004-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.