Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 250 x 10mm

Catalog No. 502396655 Shop All Phenomenex Products
Encompass

Venusil 5m ASB C18(T) 300 VS952510-T

Catalog No. 50-239-6655 Supplier Phenomenex Inc Supplier No. VS952510T
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 5m ASB C18(T) 300 VS952510-T

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.