Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 125 x 4.6mm

Catalog No. 502393457 Shop All Phenomenex Products
Encompass

Venusil 5m ASB C8 150 VS851305-0

Catalog No. 50-239-3457 Supplier Phenomenex Inc Supplier No. VS8513050
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 5m ASB C8 150 VS851305-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.