Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Venusil 5µm HILIC 100Å

Catalog No. 502412651 Shop All Phenomenex Products
Encompass

Venusil 5µm HILIC 100Å Guard Cartridge 10 x 10mm

Catalog No. 50-241-2651 Supplier Phenomenex Inc Supplier No. VH9501100S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 5µm HILIC 100Å Guard Cartridge 10 x 10mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.