Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 100 x 4.6mm

Catalog No. 502393404 Shop All Phenomenex Products
Encompass

Venusil 5m PAH 200A VP951005-L

Catalog No. 50-239-3404 Supplier Phenomenex Inc Supplier No. VP951005L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 5m PAH 200A VP951005-L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.