Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 50 x 4.6mm

Catalog No. 502394629 Shop All Phenomenex Products
Encompass

Venusil 5m PFP 120 VF950505-0

Catalog No. 50-239-4629 Supplier Phenomenex Inc Supplier No. VF9505050
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 5m PFP 120 VF950505-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.