Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Venusil 5µm XBP C18 150Å

Catalog No. 502412706 Shop All Phenomenex Products
Encompass

Guard Cartridge 10 x 21.2mm

Catalog No. 50-241-2706 Supplier Phenomenex Inc Supplier No. VX950120L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guard Cartridge 10 x 21.2mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.