Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 250 x 21.2mm

Catalog No. 502393681 Shop All Phenomenex Products
Encompass

Venusil 5m XBP C18(A) 120 VX952520-A

Catalog No. 50-239-3681 Supplier Phenomenex Inc Supplier No. VX952520A
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Venusil 5m XBP C18(A) 120 VX952520-A

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.