Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc LC Column 50 x 3mm

Catalog No. 502396047 Shop All Phenomenex Products
Encompass

Venusil 5m XBP C8(L) 150 VX850503-L

Catalog No. 50-239-6047 Supplier Phenomenex Inc Supplier No. VX850503L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 5m XBP C8(L) 150 VX850503-L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.