Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Sorbent, 10g/Vial

Catalog No. 502395262 Shop All Phenomenex Products
Encompass

Venusil 5m XBP Silica 100 VSI950010-0

Catalog No. 50-239-5262 Supplier Phenomenex Inc Supplier No. VSI9500100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Venusil 5m XBP Silica 100 VSI950010-0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.