Learn More
Phenomenex Inc Venusil AQ C18
Catalog No. 502412689 Shop All Phenomenex Products
Encompass_Preferred
3.0*150mm, 3µm, 100A
Catalog No. 50-241-2689 Supplier Phenomenex Inc Supplier No. VA9315030Q
3.0*150mm, 3µm, 100A