Learn More
Phenomenex Inc Venusil ASB C8
Catalog No. 502412833 Shop All Phenomenex Products
Encompass_Preferred
4.6*250mm, 10m, 150A
Catalog No. 50-241-2833 Supplier Phenomenex Inc Supplier No. VS8025050
Add to Cart
Add to Cart
4.6*250mm, 10m, 150A