Learn More
Phenomenex Inc Venusil XBP C18(A) Sorbent
Catalog No. 502412692 Shop All Phenomenex Products
Encompass_Preferred
Venusil XBP C18(A) Sorbent 5µm, 120A, 10g/bottle
Catalog No. 50-241-2692 Supplier Phenomenex Inc Supplier No. VX950010A
Venusil XBP C18(A) Sorbent 5µm, 120A, 10g/bottle