Learn More
Phenomenex Inc Venusil XBP C18(T) Sorbent
Catalog No. 502412882 Shop All Phenomenex Products
Encompass_Preferred
Venusil XBP C18(T) Sorbent 5µm, 300A, 10g/bottle
Catalog No. 50-241-2882 Supplier Phenomenex Inc Supplier No. VX950010T
Venusil XBP C18(T) Sorbent 5µm, 300A, 10g/bottle