Learn More
ABclonal Technology VEPH1 Rabbit pAb
Supplier: ABclonal Technology A152281000UL
Predicted to enable phosphatidylinositol-5-phosphate binding activity. Involved in negative regulation of SMAD protein signal transduction and negative regulation of transforming growth factor beta receptor signaling pathway. Predicted to be active in plasma membrane.