Learn More
TA Instruments Ver 1 Stainless Steel Double Gap Concentric Cylinder Cup, Inner Radius 20 mm, Outer Radius 22.38 mm, Use with Upper Double Wall Rotor, Sample Volume 6.48 mL, for HR X0, DHR and AR-Series Peltier Jacket
Catalog No. 5000580807 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Ver 1 Stainless Steel Double Gap Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket. It is designed to be used with the upper double wall rotor. Cup inner radius is 20.00 mm, outer radius is 22.38 mm. Sample volume with the double wall rotor is 6.48 mL.
Catalog No. 50-005-80807 Supplier TA Instruments Supplier No. 545613.901
Waters TA Instruments Ver 1 Stainless Steel Double Gap Concentric Cylinder Cup is for use with HR X0, DHR, and AR-Series Peltier Jacket. It is designed to be used with the upper double wall rotor. Cup inner radius is 20.00 mm, outer radius is 22.38 mm. Sample volume with the double wall rotor is 6.48 mL.