Learn More
Apexbio Technology LLC VER 155008 1134156-31-2 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062261
Encompass_Preferred
VER 155008 (CAS 1134156-31-2) is a small molecule inhibitor targeting members of the heat shock protein 70 (Hsp70) chaperone family It primarily inhibits Hsp70 by binding within its ATPase domain and suppressing its intrinsic ATPase activity thereby reducing chaperone-mediated cell survival functions VER 155008 demonstrates potent inhibition of Hsp70 (IC50 0 5 M) and exhibits moderate inhibitory action against related chaperones Hsc70 and glucose-regulated protein Grp78 In cancer cell models this compound diminishes cell proliferation and promotes apoptosis suggesting utility as a research tool in studying chaperone-dependent tumor cell survival pathways
Catalog No. 50-000-62261 Supplier Apexbio Technology LLC Supplier No. A43875.1
VER 155008 (CAS 1134156-31-2) is a small molecule inhibitor targeting members of the heat shock protein 70 (Hsp70) chaperone family It primarily inhibits Hsp70 by binding within its ATPase domain and suppressing its intrinsic ATPase activity thereby reducing chaperone-mediated cell survival functions VER 155008 demonstrates potent inhibition of Hsp70 (IC50 0 5 M) and exhibits moderate inhibitory action against related chaperones Hsc70 and glucose-regulated protein Grp78 In cancer cell models this compound diminishes cell proliferation and promotes apoptosis suggesting utility as a research tool in studying chaperone-dependent tumor cell survival pathways