Learn More
Medchemexpress LLC HY-10941 5mg Medchemexpress, VER-155008 CAS:1134156-31-2 Purity:>98%
SDP
Catalog No. 501968699
Encompass_Preferred
Medchemexpress, HY-10941 5mg VER-155008 CAS:1134156-31-2 VER-155008 is an inhibitor of Hsp70, with IC50s of 0.5 μM, 2.6 μM, and 2.6 μM for Hsp70, Hsc70 and Grp7, respectively, and with a Kd of 0.3 μM for Hsp70. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8699 Supplier Medchemexpress LLC Supplier No. HY109415MG
Medchemexpress, HY-10941 5mg VER-155008 CAS:1134156-31-2 VER-155008 is an inhibitor of Hsp70, with IC50s of 0.5 μM, 2.6 μM, and 2.6 μM for Hsp70, Hsc70 and Grp7, respectively, and with a Kd of 0.3 μM for Hsp70. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.