Learn More
Medchemexpress LLC HY-15986 2mg Medchemexpress, VER-49009 CAS:558640-51-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY159862MG
Encompass_Preferred
Medchemexpress, HY-15986 2mg VER-49009 CAS:558640-51-0 VER-49009 is a Hsp90 inhibitor, with an IC50 of 25 nM and a Kd of 78 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1857