Learn More
TARGETMOL CHEMICALS INC VER-82576 10MG
SDP
Catalog No. 502581030
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VER-82576 (NVP-BEP800) a synthetic HSP90(beta) inhibitor (IC50 58 nM) exhibits >70-fold selectivity against Hsp90 family members Trap-1 and Grp94. purity: 99%
Catalog No. 50-258-1030 Supplier TARGETMOL CHEMICALS INC Supplier No. T211410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. VER-82576 (NVP-BEP800) a synthetic HSP90(beta) inhibitor (IC50 58 nM) exhibits >70-fold selectivity against Hsp90 family members Trap-1 and Grp94. purity: 99%