Learn More
Medchemexpress LLC HY-10942 5mg Medchemexpress, VER-82576 CAS:847559-80-2 Purity:>98%
SDP
Catalog No. 501968700
Encompass_Preferred
Medchemexpress, HY-10942 5mg VER-82576 CAS:847559-80-2 VER-82576 (NVP-BEP800) is a potent, orally available and selective Hsp90 inhibitor, with an IC50 of 58 nM for Hsp90β; VER-82576 also slightly blocks Grp94 and Trap-1, with IC50s of 4.1 and 5.5 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8700 Supplier Medchemexpress LLC Supplier No. HY109425MG
Medchemexpress, HY-10942 5mg VER-82576 CAS:847559-80-2 VER-82576 (NVP-BEP800) is a potent, orally available and selective Hsp90 inhibitor, with an IC50 of 58 nM for Hsp90β; VER-82576 also slightly blocks Grp94 and Trap-1, with IC50s of 4.1 and 5.5 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.