Learn More
Selleck Chemical LLC VER155008 50mg 1134156-31-2 C07
SDP
Supplier:  Selleck Chemical LLC S775150MG
Encompass_Preferred
VER155008 (C07) is a potent Hsp70 family inhibitor with IC50 of 0.5 ?M, 2.6 ?M, and 2.6 ?M in cell-free assays for HSP70, HSC70, and GRP78 (HSPA5, Bip), respectively, >100-fold selectivity over HSP90. VER155008 inhibits autophagy and causes reduced levels of HSP90 client proteins. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5210