Learn More
Sino Biological VeraBIND Clean Bead Kit 1 Kit (100 Tests)
Catalog No. 5000696194
Encompass_Preferred
VeraBIND™ Clean Beads target and remove endogenous interferences from serum, plasma, saliva and saline oral rinse (SOR) samples.
Catalog No. 50-006-96194 Supplier Sino Biological Supplier No. KIT500300V1
VeraBIND™ Clean Beads target and remove endogenous interferences from serum, plasma, saliva and saline oral rinse (SOR) samples.