Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Veraguensin | 19950-55-1 | 5 MG
SDP

Catalog No. 5000419583
Encompass_Preferred

Veraguensin is a lignan compound derived from Magnolia sp., known for its ability to inhibit bone resorption.

  • Purity: 99.89%
  • Molecular weight: 372.45; Formula: C22H28O5
  • Appearance: Solid, white to off-white
  • Source: Plants (Magnolia Liliflora Desr.)
  • Shipping: Room temperature (continental US)
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month, protect from light
  • Solubility (in vitro): 25 mg/mL in DMSO
  • Solubility (in vivo): Protocols provided for various solutions

Catalog No. 50-004-19583 Supplier Medchemexpress LLC Supplier No. HYN50235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Veraguensin is a lignan compound derived from Magnolia sp., known for its ability to inhibit bone resorption.

  • Purity: 99.89%
  • Molecular weight: 372.45; Formula: C22H28O5
  • Appearance: Solid, white to off-white
  • Source: Plants (Magnolia Liliflora Desr.)
  • Shipping: Room temperature (continental US)
  • Storage (solid): 4°C, protect from light
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month, protect from light
  • Solubility (in vitro): 25 mg/mL in DMSO
  • Solubility (in vivo): Protocols provided for various solutions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.