Learn More
TARGETMOL CHEMICALS INC VERAPAMIL 200MG
SDP
Catalog No. 502581033
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Verapamil (CP-16533-1) is a calcium channel blocker and an orally active and effective inhibitor of P-gp. Verapamil inhibits CYP3A4 and can be used in studies about the treatment of high blood pressure heart arrhythmias and angina research. purity: 99%
Catalog No. 50-258-1033 Supplier TARGETMOL CHEMICALS INC Supplier No. T20656200MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Verapamil (CP-16533-1) is a calcium channel blocker and an orally active and effective inhibitor of P-gp. Verapamil inhibits CYP3A4 and can be used in studies about the treatment of high blood pressure heart arrhythmias and angina research. purity: 99%