Learn More
Medchemexpress LLC Verapamil-d3 hydrochloride | 2714485-49-9 | 99.57% | C27H36D3ClN2O4 | 1 MG
SDP
Catalog No. 5000432179
Encompass_Preferred
Verapamil-d3 hydrochloride is the deuterium labeled Verapamil hydrochloride. This compound acts as a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4 and has potential for research related to high blood pressure, heart arrhythmias, and angina.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-32179 Supplier Medchemexpress LLC Supplier No. HYA0064S1MG
Add to Cart
Add to Cart
Verapamil-d3 hydrochloride is the deuterium labeled Verapamil hydrochloride. This compound acts as a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4 and has potential for research related to high blood pressure, heart arrhythmias, and angina.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS