Learn More
Medchemexpress LLC Verapamil-d6 hydrochloride | 1185032-80-7 | C27H33D6ClN2O4 | 1 MG
SDP
Catalog No. 5000432155
Encompass_Preferred
Verapamil-d6 (CP-16533-1-d6) hydrochloride is a deuterium-labeled version of Verapamil hydrochloride. Verapamil hydrochloride is recognized as a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4 and is relevant for research into conditions such as high blood pressure, heart arrhythmias, and angina.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32155 Supplier Medchemexpress LLC Supplier No. HYA0064S11MG
Verapamil-d6 (CP-16533-1-d6) hydrochloride is a deuterium-labeled version of Verapamil hydrochloride. Verapamil hydrochloride is recognized as a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4 and is relevant for research into conditions such as high blood pressure, heart arrhythmias, and angina.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.