Learn More
Apexbio Technology LLC Verapamil HCl 152-11-4 5g
SDP
Catalog No. 5000062347
Encompass_Preferred
Verapamil HCl (CAS 152-11-4) is a small molecule belonging to the phenylalkylamine class characterized by selective inhibition of L-type calcium channels By blocking calcium influx through voltage-dependent calcium channels verapamil modulates intracellular calcium signaling affecting smooth muscle contraction cardiac electrophysiology and neurotransmitter release It is frequently utilized in research to investigate calcium channel-related physiological processes cardiovascular function and cellular signaling mechanisms
Catalog No. 50-000-62347 Supplier Apexbio Technology LLC Supplier No. B18675000
Verapamil HCl (CAS 152-11-4) is a small molecule belonging to the phenylalkylamine class characterized by selective inhibition of L-type calcium channels By blocking calcium influx through voltage-dependent calcium channels verapamil modulates intracellular calcium signaling affecting smooth muscle contraction cardiac electrophysiology and neurotransmitter release It is frequently utilized in research to investigate calcium channel-related physiological processes cardiovascular function and cellular signaling mechanisms