Learn More
AdipoGen ()-Verapamil . hydrochloride (USP Grade)
SDP
Numéro de catalogue. 501146599
Encompass_Preferred
(?)-Verapamil . hydrochloride (USP Grade), AdipoGen Life Sciences. Chemical. CAS: 152-11-4. Formula: C27H38N2O4 . HCl. MW: 454.6 . 36.5. Synthetic. alpha1-Adrenergic receptor (alpha-AR) antagonist. Prototypical calcium channel protein inhibitor that blocks the L-type Ca2+ channels in smooth and cardiac muscle cells. Vasodilator known to reduce the renal clearance of digoxin and used to control hypertension, angina, cardiac arrhythmia and vascular headaches. Apoptosis inducer in primary and metastatic colon adenocarcinoma human cell lines in vitro. Inhibitor of drug efflux pump proteins such as Mdr (P-glycoprotein) which are often over-expressed in certain tumor cell lines. Substrate of CYP3A4 and CYP2C6. Used in fluorescent cell sorting for DNA content, as it blocks efflux of a variety of DNA-binding fluorophores such as Hoechst 33342. Anti-diabetic in animal models by limiting TXNIP expression.
Numéro de catalogue. 50-114-6599 Fournisseur AdipoGen Code fournisseur AGCR13627G005
Ajouter au panier
Ajouter au panier
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
(?)-Verapamil . hydrochloride (USP Grade), AdipoGen Life Sciences. Chemical. CAS: 152-11-4. Formula: C27H38N2O4 . HCl. MW: 454.6 . 36.5. Synthetic. alpha1-Adrenergic receptor (alpha-AR) antagonist. Prototypical calcium channel protein inhibitor that blocks the L-type Ca2+ channels in smooth and cardiac muscle cells. Vasodilator known to reduce the renal clearance of digoxin and used to control hypertension, angina, cardiac arrhythmia and vascular headaches. Apoptosis inducer in primary and metastatic colon adenocarcinoma human cell lines in vitro. Inhibitor of drug efflux pump proteins such as Mdr (P-glycoprotein) which are often over-expressed in certain tumor cell lines. Substrate of CYP3A4 and CYP2C6. Used in fluorescent cell sorting for DNA content, as it blocks efflux of a variety of DNA-binding fluorophores such as Hoechst 33342. Anti-diabetic in animal models by limiting TXNIP expression.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.