Learn More
Selleck Chemical LLC Verapamil HCl 50mg 152-11-4
SDP
Supplier:  Selleck Chemical LLC S420250MG
Encompass_Preferred
Verapamil HCl is an L-type calcium channel blocker that is a class IV anti-arrhythmia agent. Verapamil inhibits both permeability-glycoprotein (P-gp) and CYP3A4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0620221