Learn More
AdipoGen Verapamil HClUSP Grade
SDP
Catalog No. 502562260
Encompass_Preferred
Chemical. CAS 152-11-4. Formula C27H38N2O4 . HCl. MW 454.6 . 36.5. Synthetic. alpha1-Adrenergic receptor alpha-AR antagonist. Prototypical calcium channel protein inhibitor that blocks the L-type Ca2 channels in smooth and cardiac muscle cells. Vasodilator known to reduce the renal clearance of digoxin and used to control hypertension, angina, cardiac arrhythmia and vascular headaches. Apoptosis inducer in primary and metastatic colon adenocarcinoma human cell lines in vitro. Inhibitor of drug efflux pump proteins such as Mdr P-glycoprotein which are often over-expressed in certain tumor cell lines. Substrate of CYP3A4 and CYP2C6. Used in fluorescent cell sorting for DNA content, as it blocks efflux of a variety of DNA-binding fluorophores such as Hoechst 33342. Anti-diabetic in animal models by limiting TXNIP expression.
Catalog No. 50-256-2260 Supplier AdipoGen Supplier No. AGCR13627G001
Chemical. CAS 152-11-4. Formula C27H38N2O4 . HCl. MW 454.6 . 36.5. Synthetic. alpha1-Adrenergic receptor alpha-AR antagonist. Prototypical calcium channel protein inhibitor that blocks the L-type Ca2 channels in smooth and cardiac muscle cells. Vasodilator known to reduce the renal clearance of digoxin and used to control hypertension, angina, cardiac arrhythmia and vascular headaches. Apoptosis inducer in primary and metastatic colon adenocarcinoma human cell lines in vitro. Inhibitor of drug efflux pump proteins such as Mdr P-glycoprotein which are often over-expressed in certain tumor cell lines. Substrate of CYP3A4 and CYP2C6. Used in fluorescent cell sorting for DNA content, as it blocks efflux of a variety of DNA-binding fluorophores such as Hoechst 33342. Anti-diabetic in animal models by limiting TXNIP expression.