Learn More
Medchemexpress LLC Verapamil hydrochloride | 152-11-4 | 491.06 | 500 MG
SDP
Catalog No. 5000365904
Encompass_Preferred
Verapamil hydrochloride is a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4 and is used in research for high blood pressure, heart arrhythmias, and angina.
  • Acts as a calcium channel blocker
  • Functions as a potent and orally active first-generation P-glycoprotein (P-gp) inhibitor
  • Inhibits CYP3A4
  • Useful for research into high blood pressure
  • Used in heart arrhythmias research
  • Applied in angina research
  • Useful for the prophylaxis of atrioventricular reentry tachycardia
  • Modulates the atrioventricular nodal response in atrial fibrillation
Catalog No. 50-003-65904 Supplier Medchemexpress LLC Supplier No. 500MGHYA0064
Verapamil hydrochloride is a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4 and is used in research for high blood pressure, heart arrhythmias, and angina.
  • Acts as a calcium channel blocker
  • Functions as a potent and orally active first-generation P-glycoprotein (P-gp) inhibitor
  • Inhibits CYP3A4
  • Useful for research into high blood pressure
  • Used in heart arrhythmias research
  • Applied in angina research
  • Useful for the prophylaxis of atrioventricular reentry tachycardia
  • Modulates the atrioventricular nodal response in atrial fibrillation