Learn More
Medchemexpress LLC Verapamil hydrochloride | 152-11-4 | 99.98% | C27H39ClN2O4 | 25 MG
SDP
Catalog No. 5000358404
Encompass_Preferred
Verapamil hydrochloride is a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4. This compound is used for research related to high blood pressure, heart arrhythmias, and angina.
  • Calcium channel blocker
  • Potent first-generation P-glycoprotein (P-gp) inhibitor
  • Inhibits CYP3A4
  • Used in research for high blood pressure and heart arrhythmias
  • Modulates atrioventricular nodal response in atrial fibrillation
  • Decreases incidence of ventricular arrhythmias
Catalog No. 50-003-58404 Supplier Medchemexpress LLC Supplier No. 25MGHYA0064R
Verapamil hydrochloride is a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4. This compound is used for research related to high blood pressure, heart arrhythmias, and angina.
  • Calcium channel blocker
  • Potent first-generation P-glycoprotein (P-gp) inhibitor
  • Inhibits CYP3A4
  • Used in research for high blood pressure and heart arrhythmias
  • Modulates atrioventricular nodal response in atrial fibrillation
  • Decreases incidence of ventricular arrhythmias