Learn More
Medchemexpress LLC HY-A0064 5g , Verapamil (hydrochloride) (±)-Verapamil hydrochlorid CAS:152-11-4 Purity:98%
SDP
Catalog No. 501492184
Encompass_Preferred
Medchemexpress, HY-A0064 5g Verapamil (hydrochloride) (±)-Verapamil hydrochlorid CAS:152-11-4 Formula:C27H39ClN2O4 Calcium channel Purity:98% Verapamil hydrochloride is an L-type calcium channel antagonist. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2184 Supplier Medchemexpress LLC Supplier No. HYA00645G
Medchemexpress, HY-A0064 5g Verapamil (hydrochloride) (±)-Verapamil hydrochlorid CAS:152-11-4 Formula:C27H39ClN2O4 Calcium channel Purity:98% Verapamil hydrochloride is an L-type calcium channel antagonist. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.