Learn More
Enzo Life Sciences (±)-Verapamil . hydrochloride (5g). CAS: 152-11-4
SDP
Catalog No. 502009530
Encompass_Preferred
Blocks slow voltage-sensitive L-type Ca2+ channels. Adrenergic antagonist. Inhibits platelet-activating factor and PMA-stimulated prostaglandin production in Kupffer cells. Induces apoptosis. In vitro resistance modifier in drug resistant human tumor cell lines. Alternative name: 5-[(3,4-Dimethoxyphenylethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropyl-valeronitrile . HCl. Purity: ≥99% (HPLC). Appearance: White crystalline powder. Solubility: Soluble in 100% ethanol (26mg/ml), methanol or water (83mg/ml). Long Term Storage: Ambient.
Catalog No. 50-200-9530 Supplier Enzo Life Sciences Supplier No. ALX550306G005
Blocks slow voltage-sensitive L-type Ca2+ channels. Adrenergic antagonist. Inhibits platelet-activating factor and PMA-stimulated prostaglandin production in Kupffer cells. Induces apoptosis. In vitro resistance modifier in drug resistant human tumor cell lines. Alternative name: 5-[(3,4-Dimethoxyphenylethyl)methylamino]-2-(3,4-dimethoxyphenyl)-2-isopropyl-valeronitrile . HCl. Purity: ≥99% (HPLC). Appearance: White crystalline powder. Solubility: Soluble in 100% ethanol (26mg/ml), methanol or water (83mg/ml). Long Term Storage: Ambient.