Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Verapamil Hydrochloride, MP Biomedicals™

Catalog No. MP219554501 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219554501 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP219554501 Supplier MP Biomedicals, Inc Supplier No. 0219554501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Functions as a calcium channel modulator, adrenoceptor antagonist, anti-arrhythmic, cardiac depressant, and coronary vasodilator. (±)-Verapamil Hydrochloride, MP Biomedicals is an α1-antagonist and calcium channel-blocker that acts by inhibit

Acts by plugging up the channels and limiting the entry of calcium into both smooth muscle cells of arteriolar walls and the cardiac muscle cells at higher doses.

  • Blocks L-type Ca2+ channels in smooth and cardiac muscle
  • Induces apoptosis of human primary and metastatic colon adenocarcinoma cells in vitro
  • Drug resistance reversal agent acting on Pgp, e.g. decrease renal tubule elimination of digoxin
  • Increases basal ATPase activity of Pgp
  • Substrate of Cyp3A4 and CYP2C6
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152-11-4
Molecular Formula C27H39ClN2O4
Molecular Weight (g/mol) 491.069
InChI Key DOQPXTMNIUCOSY-UHFFFAOYSA-N
Synonym verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
PubChem CID 62969
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
SMILES CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl

Specifications

Melting Point 140°C to 150°C (literature)
Quantity 1 g
Solubility Information Soluble in water (7g/100mL); sparingly soluble in chloroform; soluble in ethanol (26mg/mL), isopropanol, acetone, ethyl acetate; freely soluble in methanol (>100mg/mL), DMF (>100mg/mL); practically insoluble in ether.
Formula Weight 491.1
Physical Form Powder
Recommended Storage Room Temperature (15–30°C)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.