Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Verapamil Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. V011825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V011825G 25 g
V01185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V011825G Supplier TCI America Supplier No. V011825G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 152-11-4
Molecular Formula C27H39ClN2O4
Molecular Weight (g/mol) 491.069
MDL Number MFCD00055208
InChI Key DOQPXTMNIUCOSY-UHFFFAOYSA-N
Synonym verapamil hydrochloride, verapamil hcl, manidon, calcan hydrochloride, cardibeltin, cardiabeltin, cardioprotect, veroptinstada, calaptin, caveril
PubChem CID 62969
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propan-2-ylpentanenitrile;hydrochloride
SMILES CC(C)C(CCCN(C)CCC1=CC(=C(C=C1)OC)OC)(C#N)C2=CC(=C(C=C2)OC)OC.Cl

Specifications

Melting Point 145°C
Color White
pH 5.25 (0.1% aq. soln.)
Quantity 25 g
UN Number 2811
Formula Weight 491.07
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Verapamil Hydrochloride
RTECSNumber YV8320000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.