Learn More
Medchemexpress LLC Verapamil (Standard) | 52-53-9 | C27H38N2O4 | 250 MG
SDP
Catalog No. 5000355919
Encompass_Preferred
Verapamil is a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4. This analytical standard is intended for research and analytical applications, particularly in studies related to cardiovascular disease, metabolic disease, and cancer.
  • Inhibits calcium channels
  • Functions as a P-glycoprotein (P-gp) inhibitor
  • Inhibits CYP3A4
  • Suitable for research on high blood pressure
  • Applicable for studies on heart arrhythmias
  • Used in angina research
Catalog No. 50-003-55919 Supplier Medchemexpress LLC Supplier No. 250MGHY14275
Verapamil is a calcium channel blocker and a potent, orally active first-generation P-glycoprotein (P-gp) inhibitor. It also inhibits CYP3A4. This analytical standard is intended for research and analytical applications, particularly in studies related to cardiovascular disease, metabolic disease, and cancer.
  • Inhibits calcium channels
  • Functions as a P-glycoprotein (P-gp) inhibitor
  • Inhibits CYP3A4
  • Suitable for research on high blood pressure
  • Applicable for studies on heart arrhythmias
  • Used in angina research