Learn More
NEOGEN Veratox for Fumonisin, 38 tests
SDP
Supplier:  NEOGEN 700002618
Encompass_Preferred
Veratox for Fumonisin is a competitive direct ELISA that provides a quantitative analysis of fumonisin in commodities such as barley, corn, DDGS, milo, popcorn, rice, soybeans and wheat.
Catalog No. NC1835031