Learn More
Medchemexpress LLC Veratraldehyde | 120-14-9 | 166.18 | 500 G
SDP
Catalog No. 5000416551
Encompass_Preferred
Veratraldehyde is an orally active aromatic compound and antibacterial agent. It can be isolated from essential oils of plants such as peppermint and ginger. It targets the PilY1 protein and exhibits antibacterial activity against Pseudomonas aeruginosa. Additionally, Veratraldehyde has a repellent effect against mosquitoes and ticks and can be used as a flavoring agent.
  • Orally active
  • Aromatic compound
  • Antibacterial agent
  • Isolated from essential oils of plants
  • Targets PilY1 protein
  • Exhibits antibacterial activity against Pseudomonas aeruginosa
  • Repellent effect against mosquitoes and ticks
  • Can be used as a flavoring agent
Catalog No. 50-004-16551 Supplier Medchemexpress LLC Supplier No. HYN1096500G
Veratraldehyde is an orally active aromatic compound and antibacterial agent. It can be isolated from essential oils of plants such as peppermint and ginger. It targets the PilY1 protein and exhibits antibacterial activity against Pseudomonas aeruginosa. Additionally, Veratraldehyde has a repellent effect against mosquitoes and ticks and can be used as a flavoring agent.
  • Orally active
  • Aromatic compound
  • Antibacterial agent
  • Isolated from essential oils of plants
  • Targets PilY1 protein
  • Exhibits antibacterial activity against Pseudomonas aeruginosa
  • Repellent effect against mosquitoes and ticks
  • Can be used as a flavoring agent