Learn More
Selleck Chemical LLC Veratraldehyde (3.4-Dimethoxybenzaldehyde) S3214-500mg
SDP
Catalog No. 503109335
Encompass_Preferred
Veratraldehyde (3 4-dimethoxybenzaldehyde VD VAD VAld Verapamil Related Compound E Methylvanillin) a derivative of vanillin is the chemical that is found and isolated from peppermint ginger bourbon vanilla and fruits such as raspberry Veratraldehyde is widely used as a flavorant and odorant because of its pleasant woody fragrance Veratraldehyde also acts as a redox cycle agent
Catalog No. 50-310-9335 Supplier Selleck Chemical LLC Supplier No. S3214500MG
Veratraldehyde (3 4-dimethoxybenzaldehyde VD VAD VAld Verapamil Related Compound E Methylvanillin) a derivative of vanillin is the chemical that is found and isolated from peppermint ginger bourbon vanilla and fruits such as raspberry Veratraldehyde is widely used as a flavorant and odorant because of its pleasant woody fragrance Veratraldehyde also acts as a redox cycle agent