Learn More
Medchemexpress LLC Veratramine (Nsc17821; Nsc23880) | 60-70-8 | 99.9% | 409.60 g/mol | C27H39NO2 | 10 MG
SDP
Catalog No. 5000296222
Encompass_Preferred
Veratramine (standard) is an analytical standard of veratramine intended for research and analytical applications. It functions as an orally active inhibitor of the PI3K/Akt/mTOR signaling pathway and a SIGMAR1 modulator; the compound induces autophagic apoptosis, arrests the cell cycle at G0/G1, and exhibits anti-inflammatory and neuroprotective activities useful in cancer and neuropathy studies.
  • Analytical standard for research and analytical applications.
  • Orally active inhibitor of the PI3K/Akt/mTOR signaling pathway.
  • Acts as a SIGMAR1 modulator.
  • Induces autophagic apoptosis of tumor cells.
  • Inhibits epithelial-mesenchymal transition (EMT)-related proteins, reducing tumor migration.
  • Shows neuroprotective effects in neuropathy models.
Catalog No. 50-002-96222 Supplier Medchemexpress LLC Supplier No. HYN0837R10MG
Veratramine (standard) is an analytical standard of veratramine intended for research and analytical applications. It functions as an orally active inhibitor of the PI3K/Akt/mTOR signaling pathway and a SIGMAR1 modulator; the compound induces autophagic apoptosis, arrests the cell cycle at G0/G1, and exhibits anti-inflammatory and neuroprotective activities useful in cancer and neuropathy studies.
  • Analytical standard for research and analytical applications.
  • Orally active inhibitor of the PI3K/Akt/mTOR signaling pathway.
  • Acts as a SIGMAR1 modulator.
  • Induces autophagic apoptosis of tumor cells.
  • Inhibits epithelial-mesenchymal transition (EMT)-related proteins, reducing tumor migration.
  • Shows neuroprotective effects in neuropathy models.