Learn More
Apexbio Technology LLC VERATRIC-ACID-200MG
SDP
Catalog No. 5000570168
Encompass_Preferred
Veratric acid (CAS No 93-07-2) is a naturally occurring compound widely researched in the biomedical field particularly for its potential antioxidant properties Originating from various plant sources this compound exerts its effects primarily through the modulation of oxidative stress pathways It has demonstrated in vitro activity typically in the nanomolar to low micromolar range Veratric acid is frequently used in experimental settings including in vitro cell models and animal studies to assess its efficacy in mitigating oxidative damage It is also explored in drug screening processes for its potential protective effects against cellular damage Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-70168 Supplier Apexbio Technology LLC Supplier No. C6864200
Veratric acid (CAS No 93-07-2) is a naturally occurring compound widely researched in the biomedical field particularly for its potential antioxidant properties Originating from various plant sources this compound exerts its effects primarily through the modulation of oxidative stress pathways It has demonstrated in vitro activity typically in the nanomolar to low micromolar range Veratric acid is frequently used in experimental settings including in vitro cell models and animal studies to assess its efficacy in mitigating oxidative damage It is also explored in drug screening processes for its potential protective effects against cellular damage Experimental concentrations typically depend on the specific research design