Learn More
Medchemexpress LLC Veratric acid | 93-07-2 | 182.18 | 100 G
SDP
Catalog No. 5000414687
Encompass_Preferred
Veratric acid, also known as 3,4-Dimethoxybenzoic acid, is an orally active phenolic compound found in vegetables and fruits. It is recognized for its antioxidant and anti-inflammatory properties, offering protection against hypertension-associated cardiovascular remodelling. This compound has demonstrated efficacy in reducing the expression of COX-2 and mitigating levels of PGE2 and IL-6 following UVB irradiation.
  • Orally active phenolic compound
  • Derived from vegetables and fruits
  • Possesses antioxidant activities
  • Possesses anti-inflammatory activities
  • Protects against hypertension-associated cardiovascular remodelling
  • Reduces upregulated COX-2 expression
  • Reduces levels of PGE2 and IL-6 after UVB irradiation
Catalog No. 50-004-14687 Supplier Medchemexpress LLC Supplier No. HYN2007100G
Veratric acid, also known as 3,4-Dimethoxybenzoic acid, is an orally active phenolic compound found in vegetables and fruits. It is recognized for its antioxidant and anti-inflammatory properties, offering protection against hypertension-associated cardiovascular remodelling. This compound has demonstrated efficacy in reducing the expression of COX-2 and mitigating levels of PGE2 and IL-6 following UVB irradiation.
  • Orally active phenolic compound
  • Derived from vegetables and fruits
  • Possesses antioxidant activities
  • Possesses anti-inflammatory activities
  • Protects against hypertension-associated cardiovascular remodelling
  • Reduces upregulated COX-2 expression
  • Reduces levels of PGE2 and IL-6 after UVB irradiation