Learn More
Enzo Life Sciences Veratridine, (10mg), CAS Number: 71-62-5
SDP
Supplier:  Enzo Life Sciences BMLNA1250010
Encompass_Preferred
Na+ channel activator. A Na+ channel activator. Does not affect Na+-Ca2+ exchange. Veratridine acts at neurotoxin receptor site 2 and preferentially binds to activated Na+ channels causing persistent activation. It prevents inactivation and shifts activation to more negative potentials. Induces cell death in bovine chromaffin cells. Formula: C36H51NO11. MW: 673.8. CAS Number: 71-62-5. Purity: ≥98% (TLC). Appearance: White to yellow solid. Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Use/Stability: Stable for at least 1 year after receipt when stored, as supplied, at -20°C. Stock solutions are stable for up to 3 months at -20°C. Long Term Storage: -20°C
Catalog No. NC0055822