Learn More
Medchemexpress LLC Veratridine (3-Veratroylveracevine) | 71-62-5 | 5 MG
SDP
Catalog No. 5000420980
Encompass_Preferred
Veratridine is a plant neurotoxin and a voltage-gated sodium channels (VGSCs) agonist. It inhibits the peak current of Nav1.7 with an IC50 of 18.39 μM. Veratridine regulates sodium ion channels by activating them, preventing channel inactivation, and increasing sodium ion flow.
  • Acts as a voltage-gated sodium channel agonist.
  • Inhibits the peak current of Nav1.7.
  • Increases sodium ion flow.
  • Prevents channel inactivation.
Catalog No. 50-004-20980 Supplier Medchemexpress LLC Supplier No. HYN66915MG
Veratridine is a plant neurotoxin and a voltage-gated sodium channels (VGSCs) agonist. It inhibits the peak current of Nav1.7 with an IC50 of 18.39 μM. Veratridine regulates sodium ion channels by activating them, preventing channel inactivation, and increasing sodium ion flow.
  • Acts as a voltage-gated sodium channel agonist.
  • Inhibits the peak current of Nav1.7.
  • Increases sodium ion flow.
  • Prevents channel inactivation.