Learn More
Medchemexpress LLC Veratrole-d2 | 203645-56-1 | 99.7% | 100 MG
SDP
Catalog No. 5000412896
Encompass_Preferred
Veratrole-d2 is the deuterium labeled form of Veratrole (1,2-Dimethoxybenzene), a key compound widely present in plants. It attracts pollinators and is significant in plant reproduction, species differentiation, and ecological interactions. This compound can also be demethylated by cytochrome P-450. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily used as a tracer for quantitation during drug development, and has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Useful as a tracer in drug development processes
  • Potential to impact pharmacokinetic and metabolic profiles
  • Plays a role in plant reproduction and pollinator attraction
Catalog No. 50-004-12896 Supplier Medchemexpress LLC Supplier No. HYB1812S1100MG
Veratrole-d2 is the deuterium labeled form of Veratrole (1,2-Dimethoxybenzene), a key compound widely present in plants. It attracts pollinators and is significant in plant reproduction, species differentiation, and ecological interactions. This compound can also be demethylated by cytochrome P-450. The incorporation of stable heavy isotopes like deuterium into drug molecules is primarily used as a tracer for quantitation during drug development, and has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Useful as a tracer in drug development processes
  • Potential to impact pharmacokinetic and metabolic profiles
  • Plays a role in plant reproduction and pollinator attraction